Orders LinkContactUsLinkComponent
Spray chambers and brackets
Spray chambers and brackets
Replacement tubing for ketone based organic samples, spray chamber to peri pump waste line (Sturman-Masters spray chamber), per meter
Catalog # HEWL3710035300
Supplier:   Agilent
Spray chambers and brackets
Catalog # HEWL3710035300
Supplier:  Agilent
Spray chambers and brackets
Spray chambers and brackets
Replacement tubing for ketone based organic samples, spray chamber to peri pump waste line (Sturman-Masters spray chamber), per meter
Price
Inquire For Price
Log in for availability
1 * 1 ST
Product Details & Documents
These ICP-OES series are compatible with a range of accessories and supplies designed to extend the capabilities of the instruments.
Single pass glass cyclonic spray chamber for good signal to noise performance with excellent washout.
Double pass glass cyclonic spray chamber provides improved plasma stability with organic and high TDS samples.
Sturman-Masters spray chamber is inert and minimizes interferences and improves stability with high TDS samples.
Specifications
  • Description:
    Replacement tubing for ketone based organic samples, spray chamber to peri pump waste line (Sturman-Masters spray chamber), per meter
  • Seq:
    01