ContactUsLinkComponent
Flask cell culture polystyrene working volume 250ml cell growth area 175cm² rectangular angled neck with black phenolic-style cap graduated CellBIND surface sterile 1 * 50 items
Catalog # CORN3292A
Supplier:  Corning
Flask cell culture polystyrene working volume 250ml cell growth area 175cm² rectangular angled neck with black phenolic-style cap graduated CellBIND surface sterile 1 * 50 items
Catalog # CORN3292A
Supplier:  Corning
Frequently Bought Together