Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Laboratory Refrigerator LABO-288, 280 l, +2 to +20 °C 1 * 1 items
Catalog # 471-2529
Supplier:
KIRSCH PHILIPP
Expand image
Laboratory Refrigerator LABO-288, 280 l, +2 to +20 °C 1 * 1 items
Catalog # 471-2529
Supplier:
KIRSCH PHILIPP