ContactUsLinkComponent
MATRIX: HYDROCARBON OIL, DESCRIPTION: CA @ 5000 µG/G MG, P, ZN @ 1600 µG/GSIZE: 400 G. METALLO-ORGANIC METALS IN HYDROCARBON OIL. A CCURACY ENSURED BY QUALITY TESTING WITH NIST STANDARD REFERENCE MATERIALS WHEN AVAILABLE. CERTIFICATE OF ANALYSIS SUP 1 * 1 items
Catalog # PERCN9308333
Supplier:  PerkinElmer
MATRIX: HYDROCARBON OIL, DESCRIPTION: CA @ 5000 µG/G MG, P, ZN @ 1600 µG/GSIZE: 400 G. METALLO-ORGANIC METALS IN HYDROCARBON OIL. A CCURACY ENSURED BY QUALITY TESTING WITH NIST STANDARD REFERENCE MATERIALS WHEN AVAILABLE. CERTIFICATE OF ANALYSIS SUP 1 * 1 items
Catalog # PERCN9308333
Supplier:  PerkinElmer
Frequently Bought Together