Auftragserfassung
Germany
AuftragsverfolgungKontakt
 

Pallets and sumps

Mobile sump pallet, steel, type SW2, for 2×200 l drums, loading height 420 mm, painted model, loading capacity 450 kg, weight 98,5 kg, 98,5 kg, 1380×810×1040 mm
  CEMG7399
 :   CEMO
Pallets and sumps
  CEMG7399
 :  CEMO
Pallets and sumps
Pallets and sumps

Mobile sump pallet, steel, type SW2, for 2×200 l drums, loading height 420 mm, painted model, loading capacity 450 kg, weight 98,5 kg, 98,5 kg, 1380×810×1040 mm

 
1 190,00 €
   
1 * 1 ST
 

The steel sumps provide safe storage for water-polluting as well as inflammable substances.

  • All SW sump pallets are equipped with hot-dip galvanised, removable gratings
  • A wide range of models ensures the correct solution for many application
  • Sump pallets with 100 mm ground clearance permit driving under from all sides

Mobile steel sump pallets contains two steering and two fixed castors and a pushing bar for trouble-free steering.

 
  • Gewicht:
    98,5 kg
  • Bezeichnung:
    Mobile sump pallet, steel, type SW2, for 2×200 l drums, loading height 420 mm, painted model, loading capacity 450 kg, weight 98,5 kg
  • B×T×H:
    1380×810×1040 mm
Accessories

 

  •  
    Bezeichnung
    Gewicht
    B×T×H
     
     
  • CEMG7835
    Barrel support type FB3 for GRP and steel sump pallets, capacity: 3 x 60 l
    17 kg
    1200×600×400 mm
       
    1 * 1 ST - 217,00 €

 

  •  
    Bezeichnung
    Gewicht
    B×T×H
     
     
  • CEMG7741
    Barrel support stackable for GRP and steel sump pallets, capacity: 1 x 60 l
    10 kg
    630×450×530 mm
       
    1 * 1 ST - 260,00 €

 

  •  
    Bezeichnung
    Gewicht
    B×T×H
     
     
  • CEMG7586
    Barrel support type FB1 for GRP and steel sump pallets, capacity: 1 x 60 l or 1 x 200 l
    15 kg
    600×600×350 mm
       
    1 * 1 ST - 155,00 €

 

  •  
    Bezeichnung
    Gewicht
    B×T×H
     
     
  • CEMG7587
    Barrel support type FB2 for GRP and steel sump pallets, capacity 2 x 60 l or 2 x 200 l
    21 kg
    1130×600×350 mm
       
    1 * 1 ST - 188,00 €

 

  •  
    Bezeichnung
    Gewicht
    B×T×H
     
     
  • CEMG8675
    Drum pallet, PE, for 2×200 l drums
    15 kg
    940×1380×390 mm
       
    1 * 1 ST - 281,00 €