Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
ReadyCal-Kit Poly(styrene) high, 3 x 10 Vials-4.0ml, Mp 1 620 - 7 520 000 Da 1 * 1 KIT
POSSPSKITR4H-05
:
POLYMER STANDARDS SERVICE
ReadyCal-Kit Poly(styrene) high, 3 x 10 Vials-4.0ml, Mp 1 620 - 7 520 000 Da 1 * 1 KIT
POSSPSKITR4H-05
:
POLYMER STANDARDS SERVICE