ContactUsLinkComponent
Active microprocessor control for humidifying and dehumidifying (40 – 97 % rh), incl. digital indication and auto-diagnostic system ensures even more rapid reaching of set humidity and very short recovery times while avoiding condensate formation. hu 1 * 1 items
Catalog # MEMMICO105K7
Supplier:  MEMMERT
Active microprocessor control for humidifying and dehumidifying (40 – 97 % rh), incl. digital indication and auto-diagnostic system ensures even more rapid reaching of set humidity and very short recovery times while avoiding condensate formation. hu 1 * 1 items
Catalog # MEMMICO105K7
Supplier:  MEMMERT
Frequently Bought Together