ContactUsLinkComponent
Entry port (silicone), 40 mm clear diameter, for introducing connections, moisture tight, can be closed by silicone stopper, at the back, centre right not available for ico50 with active humidity control (option k7) or humidity display (option k6) 1 * 1 items
Catalog # MEMMICO50F7
Supplier:  MEMMERT
Entry port (silicone), 40 mm clear diameter, for introducing connections, moisture tight, can be closed by silicone stopper, at the back, centre right not available for ico50 with active humidity control (option k7) or humidity display (option k6) 1 * 1 items
Catalog # MEMMICO50F7
Supplier:  MEMMERT
Frequently Bought Together