ContactUsLinkComponent
grind bowl including cover CAC128, 710 ml, dishwasher-safe, stainless steel grinding bowls with convenient storage lids 1 * 1 items
Catalog # WARICAC128
Supplier:  WARING BLENDOR
grind bowl including cover CAC128, 710 ml, dishwasher-safe, stainless steel grinding bowls with convenient storage lids 1 * 1 items
Catalog # WARICAC128
Supplier:  WARING BLENDOR