ContactUsLinkComponent
Steam Formaldehyde Biological Indicatorological Indicator strip with , Geobacillus stearothermophilus Cell Line 7953. 6 mm x 30 mm Strips Individually Packaged in Glassine Pouches, 100 strips per box, 105 population level, ISO 11138-1, 11138-3 and US 1 * 1 Each
Catalog # NAMSSTSF-05
Supplier:  CROSSTEX
Steam Formaldehyde Biological Indicatorological Indicator strip with , Geobacillus stearothermophilus Cell Line 7953. 6 mm x 30 mm Strips Individually Packaged in Glassine Pouches, 100 strips per box, 105 population level, ISO 11138-1, 11138-3 and US 1 * 1 Each
Catalog # NAMSSTSF-05
Supplier:  CROSSTEX