Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
human serum albumine lyophilized 1kg 1 * 1 kg
Catalog # IDBI32500-1KG
Supplier:
SEQENS
Expand image
human serum albumine lyophilized 1kg 1 * 1 kg
Catalog # IDBI32500-1KG
Supplier:
SEQENS
Related Information
Documentation
About H2B
Product Literature
H2B Sales Brochure
Documentation