Dilatometers, Quickfit®
Dilatometer stopper with cone size NS 14/23
Catalog # BIBBSB14DL
Supplier: SciLabware
Jump to:
Product Details & Documents
PYREX®, borosilicate glass.
- Dilatometer body is graduated with an arbitrary scale
- Interchangable stopper has open top to allow weighting with steel balls etc.
For measuring the expansion of edible oils and fats over the temperature range within which melting takes place.
Ordering information: The body and stopper should be ordered separately.
Specifications
- Description:Dilatometer stopper with cone size NS 14/23
- Seq:02