Order Entry
Germany
Orders LinkContactUsLinkComponent
Flasks, round bottom, Quickfit® ground joints

Flasks, round bottom, Quickfit® ground joints

3000 ml, 34/35 NS, 249 mm
  BIBBFR3L/5S
 :   SciLabware
Flasks, round bottom, Quickfit® ground joints
  BIBBFR3L/5S
Flasks, round bottom, Quickfit® ground joints
Flasks, round bottom, Quickfit® ground joints

3000 ml, 34/35 NS, 249 mm

 
503,00 €
   
1 * 5 ST
 

PYREX®, borosilicate glass, clear.

  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes

With white, matt labelling area.

Certifications: ISO 1773, ISO 4797.

 
  • Socket size:
    34/35 NS
  • Capacity:
    3000 ml
  • Height:
    249 mm
  • Seq:
    44
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.