Orders LinkContactUsLinkComponent
Caps for tubes
Caps for tubes
Cap for 16 mm tubes, Green
Catalog # DELT305507
Supplier:   DELTALAB
Caps for tubes
Catalog # DELT305507
Supplier:  DELTALAB
Caps for tubes
Cap for 16 mm tubes, Green
Price
143,00 €
Log in for availability
1 * 2.500 ST
Product Details & Documents
Polyethylene caps for all types of polypropylene and polystyrene tubes.
Specifications
  • Colour:
    Green
  • Description:
    Cap for 16 mm tubes
  • Seq:
    15