Thermotransfer, Druck, 12 Punkte/mm (300dpi), Medienbreite (max): 108mm, Geschwindigkeit (max.): 102mm/Sek., Multi Interface (RS232, USB), Printserver (intern 10/100), EPL, ZPL, Peeler, verschiebbarer Sensor, Real Time Clock, inkl.: Kabel (USB), Netz 1 * 1 ST
Catalog # MSPP400265
Supplier: MARKET SOURCE PART PROCESS
Frequently Bought Together