0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Germany
ContactUsLinkComponent
RECOMBINANT C4 PROTEASE PROTEIN 1 * 50 µG
ABCAAB134862-50
:
Abcam
RECOMBINANT C4 PROTEASE PROTEIN 1 * 50 µG
ABCAAB134862-50
:
Abcam