Order Entry
Germany
Orders LinkContactUsLinkComponent
pH Buffers

pH Buffers

pH buffer, 3,9 M NaCl, 2,00, 250 ml, 250 mL
Catalog # METN52004100
Supplier:   Mettler - Toledo
pH Buffers
Catalog # METN52004100
Supplier:  Mettler - Toledo
CAS Number:  
pH Buffers

pH buffer, 3,9 M NaCl, 2,00, 250 ml, 250 mL

CAS Number:  
Price
46,06 €
Log in for availability
1 * 250 mL
Product Details & Documents
Specifications

Specifications

  • Pk:
    250 mL
  • Description:
    pH buffer, 3,9 M NaCl
  • Pk Info:
    250 ml
  • pH value:
    2,00

Specifications