pH Buffers
pH buffer, 3,9 M NaCl, 7,00, 250 ml, 250 mL
Catalog # METN52004106
Supplier: Mettler - Toledo
Jump to:
Product Details & Documents
Specifications
Specifications
- Pk:250 mL
- Description:pH buffer, 3,9 M NaCl
- Pk Info:250 ml
- pH value:7,00
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



