Order Entry
Germany
Orders LinkContactUsLinkComponent
Reverse frits

Reverse frits

Reverse frit with glass stopper P4, Borosilicate glass 3.3, 100 ml, 30 mm
Catalog # NEUB4322-0100-294
Reverse frits
Catalog # NEUB4322-0100-294
Reverse frits

Reverse frit with glass stopper P4, Borosilicate glass 3.3, 100 ml, 30 mm

Price
311,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

Ideal for the filtration of substances sensitive to oxidation and hydrolysis. With filter plate, one-way cock and two standard ground-nuts.

Specifications
  • Frit Ø ext.:
    30 mm
  • Frit material:
    Borosilicate glass 3.3
  • Description:
    Reverse frit with glass stopper P4
  • Seq:
    02
  • Frit volume:
    100 ml
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.