Order Entry
Germany
Orders LinkContactUsLinkComponent
Reverse frits

Reverse frits

Reverse frit with glass stopper P4, Borosilicate glass 3.3, 100 ml, 30 mm
Catalog # NEUB4322-0100-144
Reverse frits
Catalog # NEUB4322-0100-144
Reverse frits

Reverse frit with glass stopper P4, Borosilicate glass 3.3, 100 ml, 30 mm

Price
360,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

Ideal for the filtration of substances sensitive to oxidation and hydrolysis. With filter plate, one-way cock and two standard ground-nuts.

Specifications
  • Frit Ø ext.:
    30 mm
  • Frit material:
    Borosilicate glass 3.3
  • Description:
    Reverse frit with glass stopper P4
  • Seq:
    01
  • Frit volume:
    100 ml