Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
READYCAL-KIT PULLULAN HIGH, 3 X 5 VIALS-1.5ML, MP 180 - 1 220 000 DA 1 * 1 KIT
Catalog # POSSPULKITR1H-01
Supplier:
POLYMER STANDARDS SERVICE
Expand image
READYCAL-KIT PULLULAN HIGH, 3 X 5 VIALS-1.5ML, MP 180 - 1 220 000 DA 1 * 1 KIT
Catalog # POSSPULKITR1H-01
Supplier:
POLYMER STANDARDS SERVICE