Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
VERZAMELWAGEN, LAADVERM. 100KG LICHTBLAUW RAL 5012 LXBXH 670 X 380 X 830 MM 1 * 1 ST
Catalog # KAKA727286
Supplier:
KAISER PLUS KRAFT K K
Expand image
VERZAMELWAGEN, LAADVERM. 100KG LICHTBLAUW RAL 5012 LXBXH 670 X 380 X 830 MM 1 * 1 ST
Catalog # KAKA727286
Supplier:
KAISER PLUS KRAFT K K