ContactUsLinkComponent
Lid for quartz glass crucible, fits to low, medium and high form. Inner diameter 70mm, outer diameter 74mm. The durability or operat ing cycles of our quartz glass product mainly depends on applications. For application temperatures below 1050°C, nea 1 * 1 items
  TSLA75075
Lid for quartz glass crucible, fits to low, medium and high form. Inner diameter 70mm, outer diameter 74mm. The durability or operat ing cycles of our quartz glass product mainly depends on applications. For application temperatures below 1050°C, nea 1 * 1 items
  TSLA75075