Order Entry
Germany
Orders LinkContactUsLinkComponent
 

pH Buffer solutions

Buffer solution, technical grade, 9,23, Colourless, 500 mL
Catalog # HACHS11M011
Supplier:   Hach
pH Buffer solutions
Catalog # HACHS11M011
Supplier:  Hach
CAS Number:  
pH Buffer solutions
pH Buffer solutions

Buffer solution, technical grade, 9,23, Colourless, 500 mL

CAS Number:  
Price
31,50 €
Log in for availability
1 * 500 mL
Product Details & Documents

For calibration of pH meters.

Accuracy: pH is ±0,02 at 25 °C

 : Traceable to NIST.

 : Please note that the pH 4 and 10 buffers contain formaldehyde <0,1% and the pH 7 buffer contains formaldehyde 0,1 to 1%.

Specifications

Specifications

  • Pk:
    500 mL
  • Colour:
    Colourless
  • Description:
    Buffer solution, technical grade
  • pH value:
    9,23

Specifications