Order Entry
Germany
Orders LinkContactUsLinkComponent
pH Buffer solutions

pH Buffer solutions

Buffer solution, technical grade, 4,65, Colourless, 500 mL
Catalog # HACHS11M010
Supplier:   Hach
pH Buffer solutions
Catalog # HACHS11M010
Supplier:  Hach
CAS Number:  
pH Buffer solutions
pH Buffer solutions

Buffer solution, technical grade, 4,65, Colourless, 500 mL

CAS Number:  
Price
31,50 €
Log in for availability
1 * 500 mL
Product Details & Documents

For calibration of pH meters.

Accuracy: pH is ±0,02 at 25 °C

Zertifizierung: Traceable to NIST.

Achtung! Bitte beachten: Please note that the pH 4 and 10 buffers contain formaldehyde <0,1% and the pH 7 buffer contains formaldehyde 0,1 to 1%.

Specifications

Specifications

  • Pk:
    500 mL
  • Colour:
    Colourless
  • Description:
    Buffer solution, technical grade
  • pH value:
    4,65

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.