ContactUsLinkComponent
PLATELET DERIVED GROWTH FACTOR. From Outdated Human PlateletsLyophilized, partially purified.Activity: >5 x 104 units/mg protein.Uni t Definition: One unit stimulates DNA synthesis in 50% of the 3T3 cells in a confluent monolayer using a microtiter p 1 * 50 AU
Catalog # ICNA0215020450
Supplier:  MP Biomedicals
PLATELET DERIVED GROWTH FACTOR. From Outdated Human PlateletsLyophilized, partially purified.Activity: >5 x 104 units/mg protein.Uni t Definition: One unit stimulates DNA synthesis in 50% of the 3T3 cells in a confluent monolayer using a microtiter p 1 * 50 AU
Catalog # ICNA0215020450
Supplier:  MP Biomedicals