ContactUsLinkComponent
MP SILICA, 32 - 63, 60A. Mean Pore Size: 60Å Specific Surface Area: 500-600 m2 /g Particle Size: 32-63 µm Weight: 0.4-0.5 g/ml pH: 7 .0 1 * 1 kg
Catalog # ICNA0202825
Supplier:  MP Biomedicals
MP SILICA, 32 - 63, 60A. Mean Pore Size: 60Å Specific Surface Area: 500-600 m2 /g Particle Size: 32-63 µm Weight: 0.4-0.5 g/ml pH: 7 .0 1 * 1 kg
Catalog # ICNA0202825
Supplier:  MP Biomedicals
Frequently Bought Together