ContactUsLinkComponent
MP SILICA, 100 - 200 µM, 60Å. Mean Pore Size: 60Å Specific Surface Area: 500-600 m2 /g Particle Size: 100-200 µm Weight: 0.4-0.5 g/m l pH: 7.0 1 * 25 kg
Catalog # ICNA0202761
Supplier:  MP Biomedicals
MP SILICA, 100 - 200 µM, 60Å. Mean Pore Size: 60Å Specific Surface Area: 500-600 m2 /g Particle Size: 100-200 µm Weight: 0.4-0.5 g/m l pH: 7.0 1 * 25 kg
Catalog # ICNA0202761
Supplier:  MP Biomedicals
Frequently Bought Together