ContactUsLinkComponent
MP SILICA, 12-26 µM, 60Å. Mean Pore Size: 60ÅSpecific Surface Area: 500-600 m2 /gParticle Size: 12-26 µmWeight: 0.4 g/mlpH: 6.5-7.0 1 * 100 g
Catalog # ICNA0202735
Supplier:  MP Biomedicals
MP SILICA, 12-26 µM, 60Å. Mean Pore Size: 60ÅSpecific Surface Area: 500-600 m2 /gParticle Size: 12-26 µmWeight: 0.4 g/mlpH: 6.5-7.0 1 * 100 g
Catalog # ICNA0202735
Supplier:  MP Biomedicals
Frequently Bought Together