Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
CITRIC ACID ANHYDROUS. Cell Culture Reagent Anhydrous Crystalline 1 * 100 g
Catalog # ICNA0219464180
Supplier:
MP Biomedicals
Expand image
CITRIC ACID ANHYDROUS. Cell Culture Reagent Anhydrous Crystalline 1 * 100 g
Catalog # ICNA0219464180
Supplier:
MP Biomedicals