Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
SC 150-S13 (230V/50Hz) HEATING BATH CIRCULATOR STEEL, SC150-S13, Amb+13 TO +150 C, 7-11 L BATH VOLUME, 2.0 kW HEATER, PUMP 17 L/MIN & 300 MBAR USB UL/CSA, 1 * 1 items
Catalog # HAKA1531131
Supplier:
THERMO ELECTRON LED
Expand image
SC 150-S13 (230V/50Hz) HEATING BATH CIRCULATOR STEEL, SC150-S13, Amb+13 TO +150 C, 7-11 L BATH VOLUME, 2.0 kW HEATER, PUMP 17 L/MIN & 300 MBAR USB UL/CSA, 1 * 1 items
Catalog # HAKA1531131
Supplier:
THERMO ELECTRON LED