Digital Solutions
Applications
Support
About
Germany
ContactUsLinkComponent
SC 100-S21 (230V/50Hz) HEATING BATH CIRCULATOR STEEL, SC100-S21, Amb+13 to 100 C, 7-17 L BATH VOLUME, 2.0 kW HEATER, PUMP 17 L/MIN & 300 MBAR 1 * 1 items
HAKA1521211
:
THERMO ELECTRON LED
SC 100-S21 (230V/50Hz) HEATING BATH CIRCULATOR STEEL, SC100-S21, Amb+13 to 100 C, 7-17 L BATH VOLUME, 2.0 kW HEATER, PUMP 17 L/MIN & 300 MBAR 1 * 1 items
HAKA1521211
:
THERMO ELECTRON LED