Precision™ Circulating Water Baths
Circulating water bath, 89 L
HAKATSCIR89
Ensure temperature uniformity and control with these Precision™ circulating water baths, which is particularly critical when working with enzymes and in serological applications. Available in three different models, these high performance baths feature a microprocessor control and include a removable stainless steel gable cover, diffuser tray, and rubber duck.
Achieve enhanced temperature uniformity with perimeter-directed water flow
- Easily clean and maintain bath with coil-free internal design
- Optimise scheduling with auto-on and auto-off timers
- Accommodate taller labware with new hinged lid and extended height
- Help prevent bath overheating and damage with low-fluid protection
- Easily operate and monitor with icon-based graphical display
- Protect your work with audible alarms
: Not for use for IVD/MD in some countries.
- Description:Circulating water bath, 89 L
- Cat. No.:HAKATSCIR89
- Capacity:89 L
- Electrical:Global Voltage 100/115 V; 200/230 V; 50/60 Hz
- Ext. W×D×H:483×914×300 mm



