Order Entry
Germany
Orders LinkContactUsLinkComponent
Precision™ Circulating Water Baths

Precision™ Circulating Water Baths

Circulating water bath, 19 L
Catalog # HAKATSCIR19
Supplier:   THERMO ELECTRON LED
Precision™ Circulating Water Baths
Catalog # HAKATSCIR19
Precision™ Circulating Water Baths
Precision™ Circulating Water Baths

Circulating water bath, 19 L

Price
2 910,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

Ensure temperature uniformity and control with these Precision™ circulating water baths, which is particularly critical when working with enzymes and in serological applications. Available in three different models, these high performance baths feature a microprocessor control and include a removable stainless steel gable cover, diffuser tray, and rubber duck.

  • Achieve enhanced temperature uniformity with perimeter-directed water flow

  • Easily clean and maintain bath with coil-free internal design
  • Optimise scheduling with auto-on and auto-off timers
  • Accommodate taller labware with new hinged lid and extended height
  • Help prevent bath overheating and damage with low-fluid protection
  • Easily operate and monitor with icon-based graphical display
  • Protect your work with audible alarms

Ordering information: Not for use for IVD/MD in some countries.

Specifications
  • Description:
    Circulating water bath, 19 L
  • Seq:
    1
  • Cat. No.:
    HAKATSCIR19
  • Capacity:
    19 L
  • Electrical:
    Global Voltage 100/115 V; 200/230 V; 50/60 Hz
  • Ext. W×D×H:
    305×387×192 mm