Order Entry
Germany
Orders LinkContactUsLinkComponent
Microscope slides for printers

Microscope slides for printers

Clear
Catalog # KLINKP-SIL-3049
Supplier:   KLINIPATH
Microscope slides for printers
Catalog # KLINKP-SIL-3049
Supplier:  KLINIPATH
Microscope slides for printers

Clear

Price
498,00 €
Log in for availability
1 * 2.500 ST
Product Details & Documents

Microscope slides for slide printers.

  • Frosted ends
  • 90° ground edges
  • Finished clipped corners of 1 mm

Allowing for easy handling of automated slide printers and digital slide imagers in slide racks.

Specifications
  • Colour:
    Clear
  • Seq:
    08
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.