Ethanol absolute ≥99%
Steel drum, 20 L
Jump to:
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
Product Details & Documents
Synonyms: Ethyl alcohol
- CAS Number:64-17-5
- Formula:C₂H₅OH
- MDL Number:MFCD00003568
- Molecular Weight:46.07 g/mol
- Flash Pt:12 °C (closed cup)
- Storage Temperature:Ambient
- UN:1170
- ADR:3,II
Specifications
Specifications
- Pack type:Steel drum
- Pk:20 L
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



