Embedding cassettes, without lid
Light blue
Catalog # KLIN310-08S
Supplier: KLINIPATH
Jump to:
Product Details & Documents
Every cassette maintain its stability, rigidness and shape during processing of tissues.
Verpackung: Hoppers contain 75 standard cassettes and are specially packed for Thermo Fisher® cassette printers. Taped stacks contain 40 cassettes and are specially packed for Sakura® / Leica® cassette printers.
Specifications
- Colour:Light blue
- Seq:08