Embedding cassettes with disposable lid
Ochre
Catalog # FTEC110-11-10LMI
Supplier: FA TECH DIAGNOSTICS
Jump to:
Product Details & Documents
Embedded cassettes with hinged lid.
- Cassettes maintain their stability, rigidness and shape during processing of tissues
Ordering information: Available in different colours.
Packaging: Hoppers contain 80 cassettes which are specially packed for Thermo Fisher® cassette printers. Taped stacks contain 40 cassettes which are specially packed for Sakura® / Leica® cassette printers. Lids are packed separately.
Specifications
- Colour:Ochre
- Seq:84