Embedding cassettes with disposable lid
Yellow
Catalog # KLIN110-02S
Supplier: KLINIPATH
Jump to:
Product Details & Documents
Embedded cassettes with hinged lid.
- Cassettes maintain their stability, rigidness and shape during processing of tissues
Ordering information: Available in different colours.
Packaging: Hoppers contain 80 cassettes which are specially packed for Thermo Fisher® cassette printers. Taped stacks contain 40 cassettes which are specially packed for Sakura® / Leica® cassette printers. Lids are packed separately.
Specifications
- Colour:Yellow
- Seq:02