Orders LinkContactUsLinkComponent
Embedding cassettes with disposable lid
Embedding cassettes with disposable lid
Yellow
Catalog # FTEC110-02-10LMI
Supplier:   FA TECH DIAGNOSTICS
Embedding cassettes with disposable lid
Catalog # FTEC110-02-10LMI
Embedding cassettes with disposable lid
Embedding cassettes with disposable lid
Yellow
Price
106,00 €
Log in for availability
1 * 1.000 ST
Product Details & Documents
Embedded cassettes with hinged lid.
  • Cassettes maintain their stability, rigidness and shape during processing of tissues
Ordering information: Available in different colours.
Packaging: Hoppers contain 80 cassettes which are specially packed for Thermo Fisher® cassette printers. Taped stacks contain 40 cassettes which are specially packed for Sakura® / Leica® cassette printers. Lids are packed separately.
Specifications
  • Colour:
    Yellow
  • Seq:
    66