Orders LinkContactUsLinkComponent
Overalls, Secutex® pro 5/6
Overalls, Secutex® pro 5/6
White, XXL
Catalog # ZVGA30445-90
Overalls, Secutex® pro 5/6
Catalog # ZVGA30445-90
Overalls, Secutex® pro 5/6
Overalls, Secutex® pro 5/6
White, XXL
Price
101,00 €
Log in for availability
1 * 25 ST
Product Details & Documents
This particulate resistant disposable overall made from PP-spun fabric and PE laminate protects against fine dusts and splashes of aggressive, liquid chemicals. The overall comes complete with a protective flap over the zip, elasticated hood, back, sleeves and ankles.
  • Generous cut provides excellent freedom of movement, making it extremely comfortable to wear
  • Permeable to air and water vapour
  • Flexible, lint-free, abrasion resistant and anti-static material
Colour: White.
Type 5/6, EN 1149, Cat. III/CE 01
Certifications: CE 0624, Cat. III, Type 5 + 6.
Specifications
  • Clothing Size:
    XXL
  • Colour:
    White
  • Seq:
    04
  • Cat. No.:
    ZVGA30445-90
  • Anti-static:
    +
  • Garment application:
    Chemical protection|Laboratory|Industrial