ContactUsLinkComponent
solution for immobilization / coating of antibodies and antigens for ELISA, 10x stock solution 1 * 500 mL
Catalog # CAND121500
Supplier:  CANDOR BIOSCIENCE
solution for immobilization / coating of antibodies and antigens for ELISA, 10x stock solution 1 * 500 mL
Catalog # CAND121500
Supplier:  CANDOR BIOSCIENCE