ContactUsLinkComponent
BSA-Block ********************************************** Composition: Standard blocker with BSA and Tween pH-value: 7.3 ± 0.2 Preservative: contains 0.045% ProClin® 300 (v/v) Storage: shelf life (when stored unopened at -20°C): 1 year – tolerates rep 1 * 500 mL
Catalog # CAND115500
Supplier:  CANDOR BIOSCIENCE
BSA-Block ********************************************** Composition: Standard blocker with BSA and Tween pH-value: 7.3 ± 0.2 Preservative: contains 0.045% ProClin® 300 (v/v) Storage: shelf life (when stored unopened at -20°C): 1 year – tolerates rep 1 * 500 mL
Catalog # CAND115500
Supplier:  CANDOR BIOSCIENCE