ContactUsLinkComponent
dilution buffer for long-term storge of peroxidase conjugates, ready-to-use, for minimization of interference in immunoassays like ELISA 1 * 50 mL
Catalog # CAND270050
Supplier:  CANDOR BIOSCIENCE
dilution buffer for long-term storge of peroxidase conjugates, ready-to-use, for minimization of interference in immunoassays like ELISA 1 * 50 mL
Catalog # CAND270050
Supplier:  CANDOR BIOSCIENCE