Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
dilution buffer for long-term storge of peroxidase conjugates, ready-to-use 1 * 500 mL
Catalog # CAND220500
Supplier:
CANDOR BIOSCIENCE
Expand image
dilution buffer for long-term storge of peroxidase conjugates, ready-to-use 1 * 500 mL
Catalog # CAND220500
Supplier:
CANDOR BIOSCIENCE