ContactUsLinkComponent
sample and antibody diluton buffer, ready-to-use, for minimization of interference in immunoassays like ELISA 1 * 50 mL
Catalog # CAND101050
Supplier:  CANDOR BIOSCIENCE
sample and antibody diluton buffer, ready-to-use, for minimization of interference in immunoassays like ELISA 1 * 50 mL
Catalog # CAND101050
Supplier:  CANDOR BIOSCIENCE