Order Entry
Germany
Orders LinkContactUsLinkComponent
Spill control pallets, EcoPolyBlend™

Spill control pallets, EcoPolyBlend™

Spill control pallet, black, 250 liter sump capacity, for 2 drums, load capacity 1134 kg, 20,0 kg, 1245×635×457 mm
Catalog # 119-0376
Spill control pallets, EcoPolyBlend™
Catalog # 119-0376
Spill control pallets, EcoPolyBlend™
Spill control pallets, EcoPolyBlend™

Spill control pallet, black, 250 liter sump capacity, for 2 drums, load capacity 1134 kg, 20,0 kg, 1245×635×457 mm

Price
369,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

These pallets are made of up to 100% recycled polyethylene; dirt hiding black colour is 100% recycled, high visibility yellow is 35% recycled content. Fully compliant and quality engineered with a robust design, they withstand the toughest environments and are nearly impervious to chemical attack.

  • Independently validated by UL Environment
  • Seamless, robust design eliminates leaks
  • Excellent chemical compatibility and no corrosion
  • Sturdy and self-locating grates remove for easy cleaning
  • Square or space saving linear styles in multiple sizes offer storage versatility

Certifications: Meets or exceeds EPA, SPCC, NFPA 1, and the Int’l Fire Code.

Specifications
  • Weight:
    20,0 kg
  • Description:
    Spill control pallet, black, 250 liter sump capacity, for 2 drums, load capacity 1134 kg
  • Seq:
    01
  • W×D×H:
    1245×635×457 mm
Accessories

Product Information

  • Catalog #
    W×D×H
    Availability
    Price
  • Ramp, for 4 drum square
    Ramp, for 4 drum square
    119-0391
    1245 x 838 x 267 mm
    Log in for availability
    1 * 1 ST - 492,00 €