Order Entry
Germany
Orders LinkContactUsLinkComponent
GC guard columns, HydroGOLD

GC guard columns, HydroGOLD

Guard column, 0,53 mm, 10 m
Catalog # 555-9115
GC guard columns, HydroGOLD
Catalog # 555-9115
GC guard columns, HydroGOLD

Guard column, 0,53 mm, 10 m

Price
288,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

These guard columns extend analytical column lifetime by preventing degradation from harsh 'steam cleaning' water injections.

  • Highly deactivated to provide superior inertness, essential for analysis of active compounds
  • Protects against column contamination caused by non-volatile materials
Specifications
  • Column Format:
    Guard column
  • Ø int.:
    0,53 mm
  • Length:
    10 m
  • Seq:
    12
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.