Lanolin USP
Poly pail, 18 kg
Supplier: Spectrum Chemical
Jump to:
Product Details & Documents
Synonyms: Wool fat
- CAS Number:8006-54-0
- MDL Number:MFCD00081740
- Merck Index:12,05371
- Melting Pt:38…44 °C
- Density:0.932…0.945 g/cm³ (15 °C)
- Flash Pt:113 °C (closed cup)
- EINECS:232-348-6
Certificates
Search by Lot NumberSpecifications
Specifications
- Pack type:Poly pail
- Pk:18 kg



