Order Entry
Germany
Orders LinkContactUsLinkComponent
Lanolin USP

Lanolin USP

180 kg
Catalog # SPCMLA109-180KGBL
Supplier:   Spectrum Chemical
Lanolin USP
Catalog # SPCMLA109-180KGBL
Supplier:  Spectrum Chemical
CAS Number:  8006-54-0
Lanolin USP

180 kg

CAS Number:  8006-54-0
Price
Inquire For Price
Log in for availability
1 * 180 kg
Product Details & Documents
Synonyms:  Wool fat
  • CAS Number:8006-54-0
  • MDL Number:MFCD00081740
  • Merck Index:12,05371
  • Melting Pt:38…44 °C
  • Density:0.932…0.945 g/cm³ (15 °C)
  • Flash Pt:113 °C (closed cup)
  • EINECS:232-348-6

Documents

Specifications

Specifications

  • Pk:
    180 kg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.